Product Details
- SNP ID
-
rs2676080
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:33315125 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGCACTTGGGCTGAAAATCAAG[C/G]ATTGGTCCCTTGTCTTGGGTTCCTG
- Phenotype
-
MIM: 180903
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928134
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77525280] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928134
- Gene Name
- uncharacterized LOC101928134
There are no transcripts associated with this gene.
- Gene
- RYR3
- Gene Name
- ryanodine receptor 3
View Full Product Details