Product Details
- SNP ID
-
rs579909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:170226356 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTTTTCAGTGAGGGGCCTTCAGT[C/T]ATTACCTAGTGCTAGTAGTTTGGAG
- Phenotype
-
MIM: 600539
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRKCI
PubMed Links
Gene Details
- Gene
- PRKCI
- Gene Name
- protein kinase C iota
View Full Product Details