Product Details

SNP ID
rs7308
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:51355318 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGATCAATACCACCTTGACTTG[G/T]TGACCCAGAGAATTCTAATTAGATT
Phenotype
MIM: 600051
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EPS15 PubMed Links

Gene Details

Gene
EPS15
Gene Name
epidermal growth factor receptor pathway substrate 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159969.1 4106 UTR 3 NP_001153441.1
NM_001981.2 4106 UTR 3 NP_001972.1
XM_005270618.2 4106 UTR 3 XP_005270675.1
XM_017000615.1 4106 UTR 3 XP_016856104.1
XM_017000616.1 4106 UTR 3 XP_016856105.1
XM_017000617.1 4106 UTR 3 XP_016856106.1
XM_017000618.1 4106 UTR 3 XP_016856107.1
XM_017000619.1 4106 UTR 3 XP_016856108.1
XM_017000620.1 4106 UTR 3 XP_016856109.1

View Full Product Details