Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001159969.1 | Intron | NP_001153441.1 | |||
NM_001981.2 | Intron | NP_001972.1 | |||
XM_005270618.2 | Intron | XP_005270675.1 | |||
XM_017000615.1 | Intron | XP_016856104.1 | |||
XM_017000616.1 | Intron | XP_016856105.1 | |||
XM_017000617.1 | Intron | XP_016856106.1 | |||
XM_017000618.1 | Intron | XP_016856107.1 | |||
XM_017000619.1 | Intron | XP_016856108.1 | |||
XM_017000620.1 | Intron | XP_016856109.1 |