Product Details

SNP ID
rs7655290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140264448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACAGGGAAATAATAATTGTACCT[A/G]GACCTTCATACGTATAAAGATCTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCOC PubMed Links

Gene Details

Gene
SCOC
Gene Name
short coiled-coil protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001153446.1 Intron NP_001146918.1
NM_001153484.1 Intron NP_001146956.1
NM_001153552.1 Intron NP_001147024.1
NM_001153585.1 Intron NP_001147057.1
NM_001153635.1 Intron NP_001147107.1
NM_001153663.1 Intron NP_001147135.1
NM_001153690.1 Intron NP_001147162.1
NM_032547.2 Intron NP_115936.2

View Full Product Details