Product Details

SNP ID
rs11828241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1832898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTTAGACTAAAGGAGGGGGGA[C/T]GCCAGCAAAGCGGGGGCACAGGGGA
Phenotype
MIM: 607719 MIM: 191043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT8 PubMed Links

Gene Details

Gene
SYT8
Gene Name
synaptotagmin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290332.1 Intron NP_001277261.1
NM_001290333.1 Intron NP_001277262.1
NM_001290334.1 Intron NP_001277263.1
NM_138567.4 Intron NP_612634.3
XM_011520455.1 Intron XP_011518757.1
XM_011520456.2 Intron XP_011518758.1
XM_011520457.2 Intron XP_011518759.1
XM_017018528.1 Intron XP_016874017.1
XM_017018529.1 Intron XP_016874018.1
Gene
TNNI2
Gene Name
troponin I2, fast skeletal type
There are no transcripts associated with this gene.

View Full Product Details