Product Details
- SNP ID
-
rs3825175
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:121641535 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACCGCTCACTGGTTAGCCATAAGAC[C/T]GACCGACAGTTCCAGGAGCTCAACG
- Phenotype
-
MIM: 610277
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MORN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3825174] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MORN3
- Gene Name
- MORN repeat containing 3
There are no transcripts associated with this gene.
- Gene
- ORAI1
- Gene Name
- ORAI calcium release-activated calcium modulator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032790.3 |
991 |
Silent Mutation |
ACC,ACT |
T266T |
NP_116179.2 |
View Full Product Details