Product Details

SNP ID
rs7136373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121780583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGACCAGATCCTGGCTCCACTTCT[C/G]GCTAGCTGTGTGGCCCTGGGCAAGC
Phenotype
MIM: 616551
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RHOF PubMed Links

Gene Details

Gene
RHOF
Gene Name
ras homolog family member F, filopodia associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019034.2 6025 Intron NP_061907.2
Gene
TMEM120B
Gene Name
transmembrane protein 120B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080825.2 6025 UTR 3 NP_001074294.2
XM_005253843.4 6025 Intron XP_005253900.1
XM_017018853.1 6025 Intron XP_016874342.1
XM_017018854.1 6025 Intron XP_016874343.1

View Full Product Details