Product Details

SNP ID
rs3849376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:100827006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTATTGGCTACATAATAGTTAT[A/C]CTTGTTTTGGGAGTGTATCGATGGT
Phenotype
MIM: 603347
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPAS2 PubMed Links

Gene Details

Gene
NPAS2
Gene Name
neuronal PAS domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002518.3 Intron NP_002509.2
XM_005263953.2 Intron XP_005264010.1
XM_005263959.2 Intron XP_005264016.1
XM_005263960.2 Intron XP_005264017.1
XM_005263961.4 Intron XP_005264018.1
XM_011511242.2 Intron XP_011509544.1
XM_011511243.2 Intron XP_011509545.1
XM_017004214.1 Intron XP_016859703.1
XM_017004215.1 Intron XP_016859704.1
XM_017004216.1 Intron XP_016859705.1
XM_017004217.1 Intron XP_016859706.1

View Full Product Details