Product Details

SNP ID
rs6856354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155927946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAAACAAAGAAAAACTCTTGAT[A/C]ATCATTGAGCAGATATTATAATACA
Phenotype
MIM: 600550 MIM: 191070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTSO PubMed Links

Gene Details

Gene
CTSO
Gene Name
cathepsin O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001334.2 Intron NP_001325.1
Gene
TDO2
Gene Name
tryptophan 2,3-dioxygenase
There are no transcripts associated with this gene.

View Full Product Details