Product Details
- SNP ID
-
rs2884873
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:23811694 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGATGAGCCTCTGACAAGCTCA[C/G]TACTGTTGAGAAGAAGTAATGGATT
- Phenotype
-
MIM: 602916
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UBE2E1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74792451] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBE2E1
- Gene Name
- ubiquitin conjugating enzyme E2 E1
- Gene
- UBE2E1-AS1
- Gene Name
- UBE2E1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details