Product Details

SNP ID
rs11850998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23560477 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTTGAGAGAACATGTTTGTTCA[A/T]TCATTCAACAAAAATTGAGTAAACA
Phenotype
MIM: 603534 MIM: 611612
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282474.1 Intron NP_001269403.1
NM_001282475.1 Intron NP_001269404.1
NM_003917.4 Intron NP_003908.1
XM_005268167.3 Intron XP_005268224.1
XM_005268168.4 Intron XP_005268225.1
XM_005268169.3 Intron XP_005268226.1
XM_005268170.3 Intron XP_005268227.1
XM_005268172.3 Intron XP_005268229.1
XM_005268173.3 Intron XP_005268230.1
XM_005268174.3 Intron XP_005268231.1
XM_005268175.3 Intron XP_005268232.1
XM_005268177.3 Intron XP_005268234.1
XM_005268178.3 Intron XP_005268235.1
XM_005268179.3 Intron XP_005268236.1
XM_005268180.4 Intron XP_005268237.1
XM_005268182.3 Intron XP_005268239.1
XM_006720301.3 Intron XP_006720364.1
XM_011537283.2 Intron XP_011535585.1
XM_011537286.2 Intron XP_011535588.1
XM_017021740.1 Intron XP_016877229.1
XM_017021741.1 Intron XP_016877230.1
XM_017021742.1 Intron XP_016877231.1
XM_017021743.1 Intron XP_016877232.1
XM_017021744.1 Intron XP_016877233.1
XM_017021745.1 Intron XP_016877234.1
XM_017021746.1 Intron XP_016877235.1
XM_017021747.1 Intron XP_016877236.1
XM_017021748.1 Intron XP_016877237.1
Gene
JPH4
Gene Name
junctophilin 4
There are no transcripts associated with this gene.

Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

Gene
THTPA
Gene Name
thiamine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126339.3 Intron NP_001119811.1
NM_001256062.2 Intron NP_001242991.1
NM_001256321.2 Intron NP_001243250.1
NM_001256322.2 Intron NP_001243251.1
NM_001256323.2 Intron NP_001243252.1
NM_024328.5 Intron NP_077304.1
Gene
ZFHX2
Gene Name
zinc finger homeobox 2
There are no transcripts associated with this gene.

View Full Product Details