Product Details

SNP ID
rs1548082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109242666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTACAGCCTGTAGCAACTAATCAA[A/C]GAACAGAGTACACAATAAATATTTT
Phenotype
MIM: 606109
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUDCD1 PubMed Links

Gene Details

Gene
NUDCD1
Gene Name
NudC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128211.1 2106 UTR 3 NP_001121683.1
NM_032869.3 2106 UTR 3 NP_116258.2
XM_017013910.1 2106 UTR 3 XP_016869399.1
XM_017013911.1 2106 UTR 3 XP_016869400.1

View Full Product Details