Product Details
- SNP ID
-
rs2273957
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1311134 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAGGATCTCCTTGGTCAAATGAGCA[A/T]GCCTCTTCCCACCCTCAGCTTGAAG
- Phenotype
-
MIM: 604942
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FKBP1A-SDCBP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45446195] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FKBP1A-SDCBP2
- Gene Name
- FKBP1A-SDCBP2 readthrough (NMD candidate)
There are no transcripts associated with this gene.
- Gene
- SDCBP2
- Gene Name
- syndecan binding protein 2
- Gene
- SNPH
- Gene Name
- syntaphilin
There are no transcripts associated with this gene.
View Full Product Details