Product Details

SNP ID
rs11894718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:195742091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAGCACTTGTCCAATAACTGCC[A/G]GTGTATAACTCTGGTAAAGCTGGAT
Phenotype
MIM: 610061 MIM: 608733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH7 PubMed Links

Gene Details

Gene
DNAH7
Gene Name
dynein axonemal heavy chain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018897.2 Intron NP_061720.2
XM_011511487.2 Intron XP_011509789.1
XM_011511488.2 Intron XP_011509790.1
XM_011511489.2 Intron XP_011509791.1
XM_011511490.2 Intron XP_011509792.1
XM_011511491.2 Intron XP_011509793.1
XM_011511492.2 Intron XP_011509794.1
XM_011511493.2 Intron XP_011509795.1
XM_011511494.2 Intron XP_011509796.1
XM_011511495.2 Intron XP_011509797.1
XM_011511497.2 Intron XP_011509799.1
XM_017004504.1 Intron XP_016859993.1
Gene
SLC39A10
Gene Name
solute carrier family 39 member 10
There are no transcripts associated with this gene.

View Full Product Details