Product Details

SNP ID
rs4436243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97278448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGAGGCTGAGGTGGGAGGACCA[C/G]TTGAGTTGAGAGGTCAAGGCTGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100499484 PubMed Links

Gene Details

Gene
LOC100499484
Gene Name
SUGT1-1300002K09Rik pseudogene
There are no transcripts associated with this gene.

Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details