Product Details

SNP ID
rs12620793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:17883631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGGAGAAAGATAATTCCAGGC[C/T]GAGGATTCTAAGGCAGGAATGAGCA
Phenotype
MIM: 603888
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNS3 PubMed Links

Gene Details

Gene
KCNS3
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282428.1 Intron NP_001269357.1
NM_002252.4 Intron NP_002243.3
XM_011532825.1 Intron XP_011531127.1
XM_017004059.1 Intron XP_016859548.1

View Full Product Details