Product Details

SNP ID
rs17053105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52626228 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTAGGTAATAGATCATAATACAG[A/C]AAGCCAGGCATAATAAAATATTAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPA1L2 PubMed Links

Gene Details

Gene
HNRNPA1L2
Gene Name
heterogeneous nuclear ribonucleoprotein A1-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011724.1 Intron NP_001011724.1
NM_001011725.1 Intron NP_001011725.1

View Full Product Details