Product Details

SNP ID
rs2612100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:672363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTCTGGCTCTCTGCTCTGGTC[A/G]TTTTTAGACATAGTAATAGGTTGTG
Phenotype
MIM: 607427 MIM: 188350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENOSF1 PubMed Links

Gene Details

Gene
ENOSF1
Gene Name
enolase superfamily member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126123.3 3349 UTR 3 NP_001119595.1
NM_001318759.1 3349 UTR 3 NP_001305688.1
NM_001318760.1 3349 Intron NP_001305689.1
NM_017512.5 3349 UTR 3 NP_059982.2
NM_202758.3 3349 UTR 3 NP_974487.1
XM_011525679.2 3349 Intron XP_011523981.2
XM_017025833.1 3349 Intron XP_016881322.1
XM_017025834.1 3349 Intron XP_016881323.1
XM_017025835.1 3349 Intron XP_016881324.1
XM_017025836.1 3349 Intron XP_016881325.1
XM_017025837.1 3349 Intron XP_016881326.1
Gene
TYMS
Gene Name
thymidylate synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071.2 3349 Intron NP_001062.1

View Full Product Details