Product Details

SNP ID
rs17347644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101828889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCCCAAAGAGGATGACAGTTA[C/T]GTAAATTTTAAGCATTTGGTAATAT
Phenotype
MIM: 608004
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFKBIZ PubMed Links

Gene Details

Gene
NFKBIZ
Gene Name
NFKB inhibitor zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005474.2 Intron NP_001005474.1
NM_031419.3 Intron NP_113607.1
Gene
NXPE3
Gene Name
neurexophilin and PC-esterase domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134456.1 Intron NP_001127928.1
NM_145037.2 Intron NP_659474.1

View Full Product Details