Product Details

SNP ID
rs3803568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74816295 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGGGTGACGGGACTGGGGCGCC[G/A]GGGAGCCCAGGGCATGGTGAGTGTC
Phenotype
MIM: 609548
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LMAN1L PubMed Links

Gene Details

Gene
LMAN1L
Gene Name
lectin, mannose binding 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021819.2 316 Missense Mutation CAG,CGG Q105R NP_068591.2

View Full Product Details