Product Details

SNP ID
rs2298145
Assay Type
Validated
NCBI dbSNP Submissions
37
Location
Chr.1:52899432 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AAAGAAGCCAGACTCTGATCCCTGA[A/G]CACCTGGGTTTCTTGTTATTCAGCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ECHDC2 PubMed Links
Additional Information
For this assay, SNP(s) [rs12081324] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ECHDC2
Gene Name
enoyl-CoA hydratase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198961.1 Intron NP_001185890.1
NM_001198962.1 Intron NP_001185891.1
NM_001319958.1 Intron NP_001306887.1
NM_018281.3 Intron NP_060751.2
XM_011541709.2 Intron XP_011540011.1
XM_011541710.2 Intron XP_011540012.1
XM_011541711.2 Intron XP_011540013.1
XM_011541712.2 Intron XP_011540014.1
XM_011541713.2 Intron XP_011540015.1
XM_011541715.2 Intron XP_011540017.1
XM_011541719.2 Intron XP_011540021.1
XM_011541720.2 Intron XP_011540022.1
XM_011541721.2 Intron XP_011540023.1
XM_011541722.2 Intron XP_011540024.1
XM_011541723.2 Intron XP_011540025.1
XM_011541724.2 Intron XP_011540026.1
XM_011541725.2 Intron XP_011540027.1
XM_011541726.2 Intron XP_011540028.1
XM_011541727.2 Intron XP_011540029.1
XM_017001638.1 Intron XP_016857127.1
XM_017001639.1 Intron XP_016857128.1
XM_017001640.1 Intron XP_016857129.1
XM_017001641.1 Intron XP_016857130.1
XM_017001642.1 Intron XP_016857131.1
Gene
ZYG11A
Gene Name
zyg-11 family member A, cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details