Product Details

SNP ID
rs12916039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72115038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAAAAAAAATCTAAGTTTACCCA[A/G]TTTCCCTCAGGAACTATATACTACT
Phenotype
MIM: 604875 MIM: 608659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO9A PubMed Links

Gene Details

Gene
MYO9A
Gene Name
myosin IXA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006901.3 Intron NP_008832.2
XM_006720539.2 Intron XP_006720602.1
XM_011521613.2 Intron XP_011519915.1
XM_011521614.2 Intron XP_011519916.1
XM_011521615.2 Intron XP_011519917.1
XM_011521616.2 Intron XP_011519918.1
XM_011521617.2 Intron XP_011519919.1
XM_011521618.2 Intron XP_011519920.1
XM_011521619.2 Intron XP_011519921.1
XM_011521620.2 Intron XP_011519922.1
XM_011521621.2 Intron XP_011519923.1
XM_011521622.2 Intron XP_011519924.1
XM_011521623.2 Intron XP_011519925.1
XM_017022228.1 Intron XP_016877717.1
XM_017022229.1 Intron XP_016877718.1
XM_017022230.1 Intron XP_016877719.1
Gene
SENP8
Gene Name
SUMO/sentrin peptidase family member, NEDD8 specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166340.1 Intron NP_001159812.1
NM_001172109.1 Intron NP_001165580.1
NM_001172110.1 Intron NP_001165581.1
NM_001172111.1 Intron NP_001165582.1
NM_145204.3 Intron NP_660205.3
XM_005254157.3 Intron XP_005254214.1
XM_011521215.2 Intron XP_011519517.1
XM_011521217.2 Intron XP_011519519.1

View Full Product Details