Product Details

SNP ID
rs12012348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44530269 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATGGGATTACATAAGGTACTTT[C/T]ATTAGAATAAAAGCTTCCAAGTTAA
Phenotype
MIM: 300871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUNDC1 PubMed Links

Gene Details

Gene
FUNDC1
Gene Name
FUN14 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173794.3 Intron NP_776155.1

View Full Product Details