Product Details

SNP ID
rs12214600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39300960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCTGGGAAAGTTACTTCCTCAT[C/T]TGTAAAATGGAGACAGTAAGAGTCC
Phenotype
MIM: 607370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK17 PubMed Links

Gene Details

Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135111.1 Intron NP_001128583.1
NM_031460.3 Intron NP_113648.2

View Full Product Details