Product Details
- SNP ID
-
rs10123342
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:92325542 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAGCCTAAACCAAAATCCCCTAGTT[G/T]CTATTGAACACTATCGCTGTGTGTA
- Phenotype
-
MIM: 611505
MIM: 611534
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CENPP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41277649] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CENPP
- Gene Name
- centromere protein P
- Gene
- NOL8
- Gene Name
- nucleolar protein 8
View Full Product Details