Product Details

SNP ID
rs35835657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13885580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCTGGTGCCAGAGCTAGGAAC[A/G]GCTCCAAGATGGTCTCAGCCAACAG
Phenotype
MIM: 608229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024323.3 Intron NP_077299.3
XM_006722885.3 Intron XP_006722948.2
XM_011528286.2 Intron XP_011526588.1
XM_011528287.2 Intron XP_011526589.1
XM_017027300.1 Intron XP_016882789.1
XM_017027301.1 Intron XP_016882790.1
Gene
NANOS3
Gene Name
nanos C2HC-type zinc finger 3
There are no transcripts associated with this gene.

View Full Product Details