Product Details

SNP ID
rs3817541
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:50478803 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTGGAACATGGTGGAGGTGTCCCC[A/G]CTTTGCAGATAGCAAAATGGGGGCC
Phenotype
MIM: 614545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175063.5 Intron NP_778233.4
NM_206538.3 Intron NP_996261.1
XM_006723163.3 Intron XP_006723226.1
XM_011526818.2 Intron XP_011525120.1
Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
There are no transcripts associated with this gene.

View Full Product Details