Product Details

SNP ID
rs11170466
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:53192075 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGCTGGAGCATAGGGACAGATCAT[C/T]AGTTGCTCACAGTGTGTCCAGCCTG
Phenotype
MIM: 147559
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ITGB7 PubMed Links

Gene Details

Gene
ITGB7
Gene Name
integrin subunit beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000889.2 Intron NP_000880.1
XM_005268851.3 Intron XP_005268908.1
XM_005268852.4 Intron XP_005268909.1
XM_006719376.3 Intron XP_006719439.1
Gene
ZNF740
Gene Name
zinc finger protein 740
There are no transcripts associated with this gene.

View Full Product Details