Product Details

SNP ID
rs3803081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52170498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCAAGGCCTGGTGCACCAAAGCC[C/T]GGTGCTCAGGCTGGGAACAACCCGG
Phenotype
MIM: 611161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT80 PubMed Links

Gene Details

Gene
KRT80
Gene Name
keratin 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081492.1 2392 UTR 3 NP_001074961.1
NM_182507.2 2392 UTR 3 NP_872313.2
XM_005268676.3 2392 UTR 3 XP_005268733.1

View Full Product Details