Product Details

SNP ID
rs1471172
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:169096709 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGCCTAGAAAAGAAAGAGATTTTG[A/G]TTAAGGCGATTCAAAGCAGTGATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH3RF1 PubMed Links

Gene Details

Gene
SH3RF1
Gene Name
SH3 domain containing ring finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020870.3 Intron NP_065921.2

View Full Product Details