Product Details

SNP ID
rs11709691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48262179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACAGTGCAACTCCAATAAATA[A/G]GGACATTTTATTTTATTTATTTTTT
Phenotype
MIM: 616702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF589 PubMed Links

Gene Details

Gene
ZNF589
Gene Name
zinc finger protein 589
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016089.2 Intron NP_057173.2
XM_017006575.1 Intron XP_016862064.1
XM_017006576.1 Intron XP_016862065.1
XM_017006577.1 Intron XP_016862066.1
XM_017006578.1 Intron XP_016862067.1
XM_017006579.1 Intron XP_016862068.1

View Full Product Details