Product Details
- SNP ID
-
rs34162779
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:122775477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAACCGAGCATTGCAGCGGCGCT[G/T]TCCCAGCTCAGCTGGCCTTCCTCGG
- Phenotype
-
MIM: 605929
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX2
PubMed Links
Gene Details
- Gene
- SNX2
- Gene Name
- sorting nexin 2
View Full Product Details