Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002876.3 | 17 | UTR 5 | NP_002867.1 | ||
NM_058216.2 | 17 | UTR 5 | NP_478123.1 | ||
XM_006722001.3 | 17 | UTR 5 | XP_006722064.1 | ||
XM_006722002.3 | 17 | UTR 5 | XP_006722065.1 | ||
XM_006722004.3 | 17 | UTR 5 | XP_006722067.1 | ||
XM_006722005.3 | 17 | UTR 5 | XP_006722068.1 | ||
XM_011525092.2 | 17 | Intron | XP_011523394.1 | ||
XM_011525093.2 | 17 | Intron | XP_011523395.1 | ||
XM_011525094.2 | 17 | Intron | XP_011523396.1 | ||
XM_017024914.1 | 17 | UTR 5 | XP_016880403.1 | ||
XM_017024915.1 | 17 | Intron | XP_016880404.1 | ||
XM_017024916.1 | 17 | UTR 5 | XP_016880405.1 | ||
XM_017024917.1 | 17 | UTR 5 | XP_016880406.1 | ||
XM_017024918.1 | 17 | Intron | XP_016880407.1 | ||
XM_017024919.1 | 17 | Intron | XP_016880408.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001201457.1 | 17 | Intron | NP_001188386.1 | ||
NM_031272.4 | 17 | Intron | NP_112562.3 | ||
NM_198393.3 | 17 | Intron | NP_938207.2 | ||
XM_011525029.2 | 17 | Intron | XP_011523331.1 | ||
XM_017024860.1 | 17 | Intron | XP_016880349.1 | ||
XM_017024861.1 | 17 | Intron | XP_016880350.1 |