Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000857.3 | 9 | Intron | NP_000848.1 | ||
NM_001291951.1 | 9 | Intron | NP_001278880.1 | ||
NM_001291952.1 | 9 | Intron | NP_001278881.1 | ||
NM_001291953.1 | 9 | Intron | NP_001278882.1 | ||
NM_001291954.1 | 9 | Intron | NP_001278883.1 | ||
NM_001291955.1 | 9 | Intron | NP_001278884.1 | ||
XM_011531901.1 | 9 | Intron | XP_011530203.1 | ||
XM_017008130.1 | 9 | UTR 5 | XP_016863619.1 | ||
XM_017008131.1 | 9 | Intron | XP_016863620.1 | ||
XM_017008132.1 | 9 | UTR 5 | XP_016863621.1 | ||
XM_017008133.1 | 9 | Intron | XP_016863622.1 |