Product Details
- SNP ID
-
rs6461275
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:16754509 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGGTGTGCCTCTGGGTTAGCTCTT[C/G]GTACCCTCGGGATTTTAATTGAGGC
- Phenotype
-
MIM: 613139
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN13
PubMed Links
Gene Details
- Gene
- TSPAN13
- Gene Name
- tetraspanin 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014399.3 |
|
Intron |
|
|
NP_055214.1 |
View Full Product Details