Product Details

SNP ID
rs9905308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47332020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGGCAATTATGAGTACAGCTGC[A/T]ATAAACATCCAATTGTAGGTTTTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB13 PubMed Links

Gene Details

Gene
EFCAB13
Gene Name
EF-hand calcium binding domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195192.1 Intron NP_001182121.1
NM_152347.4 Intron NP_689560.3
Gene
THCAT158
Gene Name
thyroid cancer-associated transcript 158
There are no transcripts associated with this gene.

View Full Product Details