Product Details

SNP ID
rs10906094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12172261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAATAGTGAATAATCTGCTGCC[C/G]GTTAACAATTACTTAGGCCATTTTT
Phenotype
MIM: 609230
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUDT5 PubMed Links

Gene Details

Gene
NUDT5
Gene Name
nudix hydrolase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321647.1 Intron NP_001308576.1
NM_001321648.1 Intron NP_001308577.1
NM_014142.3 Intron NP_054861.2
Gene
SEC61A2
Gene Name
Sec61 translocon alpha 2 subunit
There are no transcripts associated with this gene.

View Full Product Details