Product Details

SNP ID
rs625464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118162830 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCAGGGGAGATAGTTTATTGT[C/T]GCCAGTAGACCCAAACAATAAGACA
Phenotype
MIM: 601327 MIM: 608256
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCN2B PubMed Links

Gene Details

Gene
SCN2B
Gene Name
sodium voltage-gated channel beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004588.4 4896 UTR 3 NP_004579.1
Gene
SCN4B
Gene Name
sodium voltage-gated channel beta subunit 4
There are no transcripts associated with this gene.

View Full Product Details