Product Details

SNP ID
rs7609111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:76748257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAATTTAAATAAAATCTGTGAGA[A/G]TTACACACACCTAAAGAGGAAAACG
Phenotype
MIM: 610870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRTM4 PubMed Links

Gene Details

Gene
LRRTM4
Gene Name
leucine rich repeat transmembrane neuronal 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011533117.2 Intron XP_011531419.1

View Full Product Details