Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286668.1 | 3053 | Missense Mutation | NP_001273597.1 | ||
NM_001286669.1 | 3053 | Missense Mutation | NP_001273598.1 | ||
NM_001286670.1 | 3053 | Missense Mutation | NP_001273599.1 | ||
NM_001286671.1 | 3053 | UTR 3 | NP_001273600.1 | ||
NM_001286672.1 | 3053 | Intron | NP_001273601.1 | ||
NM_001323890.1 | 3053 | Missense Mutation | NP_001310819.1 | ||
NM_001323892.1 | 3053 | UTR 3 | NP_001310821.1 | ||
NM_001323893.1 | 3053 | UTR 3 | NP_001310822.1 | ||
NM_001323899.1 | 3053 | Missense Mutation | NP_001310828.1 | ||
NM_181806.3 | 3053 | Missense Mutation | NP_861522.2 | ||
XM_017007740.1 | 3053 | Missense Mutation | XP_016863229.1 | ||
XM_017007741.1 | 3053 | Missense Mutation | XP_016863230.1 | ||
XM_017007742.1 | 3053 | Missense Mutation | XP_016863231.1 | ||
XM_017007743.1 | 3053 | UTR 3 | XP_016863232.1 | ||
XM_017007744.1 | 3053 | Missense Mutation | XP_016863233.1 |