Product Details

SNP ID
rs35543804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183920083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGACAGCAGTGACCACGCCCAC[C/T]TGAGCTACCAGCCCCACAGCACAAA
Phenotype
MIM: 605251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC5 PubMed Links

Gene Details

Gene
ABCC5
Gene Name
ATP binding cassette subfamily C member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023587.2 5832 Intron NP_001018881.1
NM_001320032.1 5832 UTR 3 NP_001306961.1
NM_005688.3 5832 UTR 3 NP_005679.2
XM_005247058.4 5832 UTR 3 XP_005247115.1
XM_005247059.4 5832 UTR 3 XP_005247116.1
XM_011512314.1 5832 UTR 3 XP_011510616.1
XM_011512315.1 5832 Intron XP_011510617.1
XM_017005492.1 5832 UTR 3 XP_016860981.1
XM_017005493.1 5832 Intron XP_016860982.1
XM_017005494.1 5832 Intron XP_016860983.1

View Full Product Details