Product Details

SNP ID
rs2274328
Assay Type
Validated
NCBI dbSNP Submissions
85
Location
Chr.1:8949385 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAACCCCTCCTTGAAGGGGCTCAAT[A/C]TGACAGGCTATGAGACCCAGGCAGG
Phenotype
MIM: 114780
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CA6 PubMed Links
Additional Information
For this assay, SNP(s) [rs58800854] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CA6
Gene Name
carbonic anhydrase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001215.3 303 Missense Mutation ATG,CTG M68L NP_001206.2
NM_001270500.1 303 Missense Mutation ATG,CTG M68L NP_001257429.1
NM_001270501.1 303 Intron NP_001257430.1
NM_001270502.1 303 Intron NP_001257431.1
XM_011542083.2 303 Missense Mutation ATG,CTG M72L XP_011540385.1
XM_011542084.2 303 Missense Mutation ATG,CTG M72L XP_011540386.1

View Full Product Details