Product Details

SNP ID
rs4413407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:104472204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTGAAGAAAGCCCTATAC[A/G]TAAAATGAAAATAATTTCTGGATAT
Phenotype
MIM: 611645
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXXC4 PubMed Links

Gene Details

Gene
CXXC4
Gene Name
CXXC finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025212.3 1822 UTR 3 NP_079488.2
XM_011532284.2 1822 UTR 3 XP_011530586.1
XM_017008652.1 1822 UTR 3 XP_016864141.1

View Full Product Details