Product Details

SNP ID
rs11715248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:140952444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGGCATCAGCCACCATGCCTGG[C/T]CAGTTTAGATTCTCATGTGTTTTAC
Phenotype
MIM: 616149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A36 PubMed Links

Gene Details

Gene
SLC25A36
Gene Name
solute carrier family 25 member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104647.1 Intron NP_001098117.1
NM_018155.2 Intron NP_060625.2
XM_005247575.4 Intron XP_005247632.1

View Full Product Details