Product Details
- SNP ID
-
rs9525471
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:41312432 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GTGCCAGCGTCCCTGGTAGCTTTCC[A/G]GGTTCTGTTCTTCCAAGGAGAAGAG
- Phenotype
-
MIM: 604601
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MTRF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2483099] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MTRF1
- Gene Name
- mitochondrial translational release factor 1
- Gene
- NAA16
- Gene Name
- N(alpha)-acetyltransferase 16, NatA auxiliary subunit
View Full Product Details