Product Details

SNP ID
rs11807810
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:206473142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTTACAGGTCAGGCCACCAGC[A/G]GCCACCCTGTGCCAGGAATGGAATC
Phenotype
MIM: 605048 MIM: 606524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKBKE PubMed Links

Gene Details

Gene
IKBKE
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193321.1 Intron NP_001180250.1
NM_001193322.1 Intron NP_001180251.1
NM_014002.3 Intron NP_054721.1
XM_005273356.2 Intron XP_005273413.1
XM_017002868.1 Intron XP_016858357.1
Gene
MIR6769B
Gene Name
microRNA 6769b
There are no transcripts associated with this gene.

Gene
SRGAP2
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
There are no transcripts associated with this gene.

View Full Product Details