Product Details

SNP ID
rs7999177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19829751 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGTATTGTAGGTGCACACCACCA[C/T]GCCCAGCACTTACATAACTTTTCTA
Phenotype
MIM: 616443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZMYM5 PubMed Links

Gene Details

Gene
ZMYM5
Gene Name
zinc finger MYM-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039649.2 Intron NP_001034738.1
NM_001039650.2 Intron NP_001034739.1
NM_001142684.1 Intron NP_001136156.1
XM_005266593.3 Intron XP_005266650.1
XM_005266594.2 Intron XP_005266651.1
XM_006719894.2 Intron XP_006719957.1
XM_011535309.1 Intron XP_011533611.1
XM_011535310.1 Intron XP_011533612.1
XM_017020849.1 Intron XP_016876338.1
XM_017020850.1 Intron XP_016876339.1
XM_017020851.1 Intron XP_016876340.1

View Full Product Details