Product Details
- SNP ID
-
rs7326471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:41463799 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGCCCCATGAGTTATGCTGAGT[C/T]TTTCCTGAATCTTTGCTGGATTCCT
- Phenotype
-
MIM: 610077
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RGCC
PubMed Links
Gene Details
- Gene
- RGCC
- Gene Name
- regulator of cell cycle
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014059.2 |
|
Intron |
|
|
NP_054778.2 |
View Full Product Details