Product Details

SNP ID
rs4846054
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:11809173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATACCTGGCCATTTTCTTCTGTTT[C/T]GTAAGTATTTGTTTTCTTGTACATA
Phenotype
MIM: 602726 MIM: 607093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCN6 PubMed Links

Gene Details

Gene
CLCN6
Gene Name
chloride voltage-gated channel 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256959.1 Intron NP_001243888.1
NM_001286.3 Intron NP_001277.1
Gene
MTHFR
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
There are no transcripts associated with this gene.

View Full Product Details